Learn More
Hamilton™ 1 mL 1000 Series Gas Tight Syringe
Description
- This syringe series boasts a variety of termination and needle options, providing exceptional versatility and adaptability.
- Dispenses volumes up to 1 mL.
Specifications
Specifications
| Esterilizables en autoclave | Not Autoclavable |
| Material | Borosilicate Glass |
| Calibre de aguja | 22 G |
| Longitud de la aguja (imperial) | 2 in |
| Tipo de producto | Gas Tight Syringe |
| Temperatura máx. (imperial) | 239 °F |
| Temperatura máxima | 115°C |
| Temperatura mín. (imperial) | 50 °F |
| Modelo | 1001 Luer Lock (TLL) |
| Temperatura mín. (métrico) | 10 °C |
| Show More |