Learn More
1,2-Bis[(2S,5S)-2,5-diisopropil-1-fosfolanil]etano, 97 +%, Thermo Scientific Chemicals
Descripción
It is employed as DuPhos and BPE ligands. It is a highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Se emplea como ligandos DuPhos y BPE. Es un ligando privilegiado altamente eficaz.
Solubilidad
Insoluble en agua.
Notas
Sensible al aire. Mantener alejado de agentes oxidantes fuertes.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]ethane |
| Punto de fusión | 67°C to 72°C |
| Cantidad | 250 mg |
| Sensibilidad | Air Sensitive |
| Información de solubilidad | Insoluble in water. |
| Formula Weight (peso de la fórmula) | 370.53 |
| Porcentaje de pureza | ≥97% |