missing translation for 'onlineSavingsMsg'
Learn More

RhoF Antibody, CoraFluor™ 1, Novus Biologicals™

Product Code. 30142065
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30142065 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30142065

Brand: Novus Biologicals (Bio-Techne) NBP298649CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RhoF Polyclonal antibody specifically detects RhoF in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno RhoF
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Polyclonal
Conjugado CoraFluor 1
Formulación PBS
Alias de gen FLJ20247, ras homolog gene family, member F (in filopodia), Rho family GTPase Rif, Rho in filopodia, rho-related GTP-binding protein RhoF, RIFARHF
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human RhoF fragment.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 54509
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.