Learn More
p97/VCP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Marca:  Novus Biologicals NB100-1557
Código de producto. 18725573
  • 645.00€ / 0.10 ml
Descripción
p97/VCP Polyclonal specifically detects p97/VCP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Western Blot 1:5000-1:15000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 1-4 ug/mg of lysate, Immunohistochemistry-Paraffin 1:200- 1:1000
15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog
0.1 mL
RUO
Certificados
Promociones
Promociones
For Research Use Only