missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7 Antibody (PCRP-HDAC7-1B6) [Janelia Fluor™ 635], Novus Biologicals™
Description
HDAC7 Monoclonal antibody specifically detects HDAC7 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antígeno | HDAC7 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | PCRP-HDAC7-1B6 |
| Conjugado | Janelia Fluor 635 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant full-length human HDAC7 protein |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?