Learn More
CRISPR-Cpf1 Antibody (2D5-6G11), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP2-78819MFV610
Código de producto. 18487946
  • ONLINE EXCLUSIVE PRICE
    640.00€
    604.80€ / 0.10 ml
    Ahorro 35.20€
    5% Descuento
Descripción
CRISPR-Cpf1 Monoclonal antibody specifically detects CRISPR-Cpf1 in Bacteria, Bacteria samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
CRISPR-Cpf1
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Store at 4C in the dark.
IgG1 κ
Western Blot
2D5-6G11
Western Blot
Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1
Partial recombinant protein made to an N-terminal portion of the Lachnospiraceae bacterium CRISPR-Cpf1 protein
0.1 mL
Cell Biology, DNA Repair, Virology Bacteria and Parasites
Bacteria
Purified
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones