missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 Antibody (2600G), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals FAB1971P
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CCR7 Monoclonal antibody specifically detects CCR7 in Human samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
| CCR7 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 2600G | |
| Flow Cytometry, CyTOF-ready | |
| BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| Synthetic peptide containing human CCR7, Accession # P32248 | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1236 | |
| Store at 4°C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido