missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetamide-d4, CDN

Product Code. 30309446
Change view
Click to view available options
Cantidad:
0.05 g
0.1 g
Product Code. Cantidad
30309446 0.1 g
30317832 0.05 g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30309446

Brand: CDN CDND59610.1G

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Nombre del producto químico o material Iodoacetamide-D4
CAS 1219802-64-8
Fórmula molecular C2 D4 I N O
Cantidad 0.1 g
Sinónimo 2-Iodoacetamide-N,2,2-d3 (ACI), 2-Iodoacetamide-N,N,2,2-d4, 2-Iodoacetamide-d4, Iodoacetamide-d4, Iodoacetamide-D4
Fórmula InChI InChI=1 S/C2H4INO/c3-1-2(4)5/h1H2,(H2,4,5)/i1D2/hD2
SMILES [2 H]N([2 H])C(=O)C([2 H])([2 H])I
Nombre IUPAC N,N,2,2-tetradeuterio-2-iodoacetamide
Peso molecular (g/mol) 188.9884
Formula Weight (peso de la fórmula) 188.9589 g/mol
Porcentaje de pureza 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.