siRNA Details
| Gene Symbol: Hint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109607.1 | NP_001103077.1 | 1, 2 | 176 | ||
| GenBank mRNA | BC168732.1 | - | 1, 2 | 189 | 6888300 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hint1| Gene Symbol: Hint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109607.1 | NP_001103077.1 | 1, 2 | 176 | ||
| GenBank mRNA | BC168732.1 | - | 1, 2 | 189 | 6888300 | |