siRNA Details
| Gene Symbol: Fam3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198771.1 | NP_942066.1 | 9 | 696 | ||
| GenBank mRNA | AY228475.1 | - | 9 | 696 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam3c| Gene Symbol: Fam3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198771.1 | NP_942066.1 | 9 | 696 | ||
| GenBank mRNA | AY228475.1 | - | 9 | 696 | ||