siRNA Details
| Gene Symbol: Mphosph8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017375.2 | NP_001017375.2 | 12 | 2468 | ||
| GenBank mRNA | BC099116.1 | - | 12 | 2411 | 7462184 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mphosph8| Gene Symbol: Mphosph8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017375.2 | NP_001017375.2 | 12 | 2468 | ||
| GenBank mRNA | BC099116.1 | - | 12 | 2411 | 7462184 | |