siRNA Details
| Gene Symbol: LINC00548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033877.1 | 6 | 2191 | |||
| GenBank mRNA | AK124383.1 | - | 6 | 2191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00548| Gene Symbol: LINC00548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033877.1 | 6 | 2191 | |||
| GenBank mRNA | AK124383.1 | - | 6 | 2191 | ||