siRNA Details
| Gene Symbol: C1orf220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033186.1 | 3 | 2470 | |||
| GenBank mRNA | AK092849.1 | - | 3 | 2470 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf220| Gene Symbol: C1orf220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033186.1 | 3 | 2470 | |||
| GenBank mRNA | AK092849.1 | - | 3 | 2470 | ||