siRNA Details
| Gene Symbol: OR5AU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004731.1 | NP_001004731.1 | 1 | 587 | ||
| GenBank mRNA | BC137304.1 | - | 1 | 625 | 9021301 | |
| BC137305.1 | - | 1 | 625 | 9021302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5AU1| Gene Symbol: OR5AU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004731.1 | NP_001004731.1 | 1 | 587 | ||
| GenBank mRNA | BC137304.1 | - | 1 | 625 | 9021301 | |
| BC137305.1 | - | 1 | 625 | 9021302 | ||