siRNA Details
| Gene Symbol: LOC102724279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302493.1 | NP_001289422.1 | 2 | 151 | ||
| GenBank mRNA | AI436266.1 | - | 2 | 299 | ||
| BX095604.1 | - | 2 | 151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724279| Gene Symbol: LOC102724279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302493.1 | NP_001289422.1 | 2 | 151 | ||
| GenBank mRNA | AI436266.1 | - | 2 | 299 | ||
| BX095604.1 | - | 2 | 151 | |||