siRNA Details
| Gene Symbol: Ap2m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053837.1 | NP_446289.1 | 3 | 449 | ||
| GenBank mRNA | BC087724.1 | - | 456 | 7320615 | ||
| M23674.1 | - | 3 | 449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap2m1| Gene Symbol: Ap2m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053837.1 | NP_446289.1 | 3 | 449 | ||
| GenBank mRNA | BC087724.1 | - | 456 | 7320615 | ||
| M23674.1 | - | 3 | 449 | |||