siRNA Details
| Gene Symbol: UFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016406.3 | NP_057490.2 | 2, 3 | 442 | ||
| XM_005245254.1 | XP_005245311.1 | 2, 3 | 394 | |||
| XM_017001450.1 | XP_016856939.1 | 3, 4 | 287 | |||
| XM_017001451.1 | XP_016856940.1 | 3, 4 | 551 | |||
| GenBank mRNA | AB154405.1 | - | 2, 3 | 188 | ||
| AF151884.1 | - | 2, 3 | 409 | |||
| AF161504.1 | - | 2, 3 | 428 | |||
| AK292776.1 | - | 2, 3 | 394 | |||
| BC005187.1 | - | 2, 3 | 434 | 3961569 | ||
| CF128121.1 | - | 2, 3 | 441 | |||

UFC1
Genomic Map