| | | Polyethylene |
| Polypropylene | Polycarbonate | LD | HD |
| Chemical | 20°C | 60°C | 20°C | 60°C | 20°C | 60°C | 20°C | 60°C |
| Acetic acid solution <90% | R | R | R | R | R | R | R | R |
| Acetic acid, glacial | R | U | U | U | A | U | R | L |
| Acetone | R | R | U | U | E | U | R | R |
| Ammonia | R | R | U | U | R | R | R | R |
| Aniline | R | R | U | U | U | U | R | R |
| Benzene | A | U | U | U | U | U | U | U |
| Boric acid | R | R | R | R | R | R | R | R |
| Bromine | U | U | U | U | U | U | U | U |
| Carbon tetrachloride | U | U | U | U | U | U | A | U |
| Chlorinated solvents | U | U | U | U | U | U | U | U |
| Chloroform | U | U | U | U | U | U | U | U |
| Cresols | U | U | U | U | U | U | U | U |
| Diethyl ether | U | U | U | U | U | U | U | U |
| Diethylene glycol | R | R | R | U | R | R | R | E |
| Ethers | U | U | A | U | E | U | A | U |
| Ethyl acetate | A | A | | | A | U | A | U |
| Ethanol | R | R | R | R | R | U | R | R |
| Ethylene glycol | R | R | R | U | R | R | E | E |
| Formaldehyde solutions 40% | R | R | R | R | R | E | R | R |
| Formic acid | R | R | R | R | R | E | R | R |
| Hydrochloric acid conc. | E | E | L | U | R | R | SW | SW |
| Hydrofluoric acid 1-60% | E | E | 20% | U | R | R | R | L |
| Hydrofluoric acid conc. | E | E | L | U | R | U | R | L |
| Hydrogen peroxide 30-90% | R | R | R | R | R | U | R | A |
| Hypochlorous acid | R | R | | | A | U | R | R |
| Magnesium hydroxide | R | R | | | R | R | R | R |
| Maleic acid | R | R | | | R | R | R | R |
| Mineral oil | R | R | R | R | A | U | R | E |
| Nitric acid <25% | R | R | R | U | R | R | R | R |
| Nitric acid 25-70% | R | E | U | U | A | U | L | U |
| Nitric acid >70 | E | U | U | U | U | U | U | U |
| Oxalic acid | R | A | R | R | R | R | R | R |
| Paraffin | R | A | R | R | A | U | | |
| Petroleum ether | A | U | R | R | U | U | U | U |
| Phosphoric acid <30% | R | R | R | R | R | R | R | R |
| Phosphoric acid 30-85% | R | R | R | R | R | A | R | R |
| Phosphoric acid >85% | R | R | R | R | A | U | R | A |
| Potasium hydroxide | 50% | 10% | U | U | R | R | R | 30% |
| Salicylic acid | R | R | R | R | R | R | R | R |
| Sodium hydroxide solution | 50% | 10% | U | U | R | R | R | 30% |
| Sulfuric acid <60% | R | R | R | R | R | R | R | R |
| Sulfuric acid 60-70% | R | U | R | R | R | A | R | A |
| Sulfuric acid >70% | E | U | U | U | A | U | A | L |
| Silicone oil | R | R | R | R | A | U | E | E |
| Stearic acid | R | R | R | R | R | R | R | R |
| Toluene | U | U | U | U | U | U | U | U |
| Trichloroethylene | U | U | U | U | U | U | V | U |
| Water | R | R | R | R | R | R | R | R |