CAS RN 81846-19-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ Treprostinil
Potent prostacyclin (PGI2) analog
2
Treprostinil, MedChemExpress
MedChemExpress Treprostinil (UT-15) is a potent DP1 and EP2 agonist with EC50 values of 0.6±0.1 and 6.2±1.2 nM, respectively.
3