CAS RN 571170-77-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ Laropiprant
Potent and selective prostaglandin D2 receptor antagonist
2
Laropiprant, MedChemExpress
MedChemExpress Laropiprant is a potent and selective DP receptor antagonist with Ki values of 0.57 nM and 2.95 nM for DP receptor and TP Receptor, respectively.