Learn More
Tocris Bioscience™ Laropiprant
Descripción
Laropiprant is a potent and selective prostaglandin D2 receptor antagonist (IC50 = 0.09 nM in washed platelets; Ki = 0.57). It exhibits selectivity for binding to PGD2 receptors over other prostanoid receptors (Ki values are ≥ 2.95 nM for TP, EP1, EP2, EP3, EP4, FP and IP). Laropiprant promotes coagulation of whole blood ex vivo and reduces bleeding time in mice in vivo. In a mouse model of intracerebral hemorrhage, Laropiprant reduces lesion volume and attenuates neurological deficit scores. The compound also reduces Niacin (Cat. No. 4106) induced vasodilation.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (3 R)-4-[(4-Chlorophenyl)methyl]-7-fluoro-1,2,3,4-tetrahydro-5-(methylsulfonyl)cyclopent[b]indole-3-acetic acid |
| CAS | 571170-77-9 |
| Cantidad | 10 mg |
| Objetivo | Prostanoid Receptor Antagonists |
| Fórmula molecular | C21H19ClFNO4S |
| Pureza | 0.98 |