CAS RN 507475-17-4
IUPAC Nombre:
2-(carbamoylamino)-5-(4-fluorophenyl)thiophene-3-carboxamide
Sinónimos:
2-(carbamoylamino)-5-(4-fluorophenyl)thiophene-3-carboxamide
Peso molecular (g/mol):
279.29
Fórmula molecular:
C12H10FN3O2S
InChi Key:
SAYGKHKXGCPTLX-UHFFFAOYSA-N
SMILES:
NC(=O)NC1=C(C=C(S1)C1=CC=C(F)C=C1)C(N)=O
1
–
2
de
2
resultados
2
TPCA-1, MedChemExpress
MedChemExpress TPCA-1 is a potent and selective inhibitor of IKK-2 with IC50 of 17.9 nM. TPCA-1 is an effective inhibitor of STAT3 phosphorylation, DNA binding, and transactivation.