CAS RN 33985-68-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
DIM-C-pPhOCH3, MedChemExpress
MedChemExpress DIM-C-pPhOCH3 is a Nur77 agonist. Nerve growth factor-induced Bα (NGFI-Bα, Nur77) is an orphan nuclear receptor.
2
DIM-C-pPhOCH3
Nur77 agonist
3
4