CAS RN 2244989-34-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ B 973B
Positive allosteric modulator of α7 nAChRs; analgesic
2
(-)-(S)-B-973B, MedChemExpress
MedChemExpress (-)-(S)-B-973B is a potent allosteric agonist and positive allosteric modulator of α7 nAChR, with antinociceptive activity.