Learn More
Tocris Bioscience™ B 973B
Descripción
B 973B is a positive allosteric modulator of α7 nAChRs (EC50 ∽0.3 μM); active enantiomer. Increases potency of acetylcholine (EC50 values are 0.007 mM and 0.3 mM in the presence and absence of B 937B, respectively). Potentiates peak acetylcholine-induced currents 6-fold relative to maximal acetylcholine, and slows channel desensitization, resulting in a 6900-fold increase in charge transfer. Decreases paw edema in vivo. Analgesic.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3,4-Difluoro-N-[(1S)-1-[6-[4-(2-pyridinyl)-1-piperazinyl]-2-pyrazinyl]ethyl]benzenepropanamide |
| CAS | 2244989-34-0 |
| Cantidad | 10 mg |
| Fórmula molecular | C24H26F2N6O |
| Pureza | 98% |