CAS RN 1799711-21-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
2
Tocris Bioscience™ dBET1
(+)-JQ1 based Degrader (PROTAC™) targeting BET bromodomains, active in vivo
3
dBET1, MedChemExpress
MedChemExpress dBET1 is a PROTAC connected by ligands for Cereblon and BRD4 with an EC50 of 430 nM. dBET1 is a PROTAC that composes of (+)-JQ1 (HY-13030) linked to NSC 527179 (HY-14658) with a linker.