Learn More
Tocris Bioscience™ dBET1
Descripción
Degrader (PROTAC®) comprising BET bromodomain antagonist (+)-JQ1 (Cat.No. 4499) conjugated to a cereblon E3 ubiquitin ligase ligand. Depletes BET bromodomains in cancer cell lines in vitro (EC50 = 430 nM in breast cancer cells) and induces apoptosis. Delays tumor growth and downregulates MYC in mice bearing human AML xenografts.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (6S)-4-(4-Chlorophenyl)-N-[4-[[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetyl]amino]butyl]-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetamide |
| CAS | 1799711-21-9 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 5 mg |
| Fórmula molecular | C38H37ClN8O7S |
| Formula Weight (peso de la fórmula) | Observed MW: 785.27 |