CAS RN 170151-24-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ Nepicastat hydrochloride
Potent and selective dopamine β-hydroxylase (DBH) inhibitor; orally bioavailable
2
Nepicastat hydrochloride, MedChemExpress
MedChemExpress Nepicastat hydrochloride (SYN-117 hydrochloride) is a selective, potent, and orally active inhibitor of dopamine-beta-hydroxylase. Nepicastat hydrochloride produces concentration-dependent inhibition of bovine (IC50=8.5 nM) and human (IC50=9 nM) dopamine-beta-hydroxylase. Nepicastat hydrochloride can cross the blood-brain barrier (BBB).
3
Tocris Bioscience™ Nepicastat hydrochloride
Potent and selective dopamine β-hydroxylase (DBH) inhibitor; orally bioavailable
4
Tocris Bioscience™ Nepicastat hydrochloride
Potent and selective dopamine β-hydroxylase (DBH) inhibitor; orally bioavailable