CAS RN 1675821-32-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ EML 425
Reversible and non-competitive CBP/p300 inhibitor
2
EML 425, MedChemExpress
MedChemExpress EML425 is a potent and selective CREB binding protein (CBP)/p300 inhibitor with IC50s of 2.9 and 1.1 μM, respectively.
3
4