Learn More
Tocris Bioscience™ EML 425
GREENER_CHOICE
Reversible and non-competitive CBP/p300 inhibitor
Marca:  Tocris Bioscience™ 5646/10
Encompass
Código de producto. 15797006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
EML 425 is a reversible and non-competitive CBP/p300 inhibitor (IC50 values are 1.1 and 2.9 μM, respectively). Cell permeable.
Especificaciones
Especificaciones
5-[(4-Hydroxy-2,6-dimethylphenyl)methylene]-1,3-bis(phenylmethyl)-2,4,6(1H,3H,5H)-pyrimidinetrione
1675821-32-5