CAS RN 1346704-33-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
5
de
5
resultados
1
Tocris Bioscience™ GSK 343
Potent and selective SAM-competitive EZH2 inhibitor
2
Tocris Bioscience™ GSK 343
Potent and selective SAM-competitive EZH2 inhibitor
3
GSK343, MedChemExpress
MedChemExpress GSK343 is a highly potent and selective EZH2 inhibitor with an IC50 of 4 nM.
4
5