Learn More
Tocris Bioscience™ GSK 343
Descripción
GSK 343 is a potent and selective SAM-competitive EZH2 inhibitor (IC50 = 4 nM), that exhibits >60 fold selectivity for EZH2 over EZH1 and a range of other methyltransferases. GSK 343 decreases H3K27me3 levels in breast cancer cells and inhibits proliferation of prostate cancer cell lines in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[(1,2-Dihydro-6-methyl-2-oxo-4-propyl-3-pyridinyl)methyl]-1-(1-methylethyl)-6-[2-(4-methyl-1-piperazinyl)-4-pyridinyl]-1H-indazole-4-carboxamide |
| CAS | 1346704-33-3 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 10 mg |
| Fórmula molecular | C31H39N7O2 |
| Formula Weight (peso de la fórmula) | Observed MW: 541.69 |