missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SLC6A19 GST (N-Term) Protein

Product Code. 18592831 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
2 μg
Unit Size:
2 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18592831 2 μg 2 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18592831 Supplier Novus Biologicals™ Supplier No. H00340024P012ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

Para utilizar con (aplicación) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulación 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID de gen (Entrez) 340024
Peso molecular MolecularWeight-theroretical: 97.5 kDa
Cantidad 2 μg
Requisitos de almacenamiento Store at -80°C. Avoid freeze/thaw cycles.
Alias de gen B0AT1, FLJ20680, FLJ34635, HND, sodium-dependent amino acid transporter system B0, sodium-dependent neutral amino acid transporter B(0)AT1, solute carrier family 6 (neurotransmitter transporter), member 19, solute carrier family 6 (neutral amino acid transporter), member 19, Solute carrier family 6 member 19, System B(0) neutral amino acid transporter AT1, system B0 neutral amino acid transporter
Símbolo de gen SLC6A19
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión SLC6A19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.