Learn More
Novus Biologicals™ Mycobacterium Tuberculosis ESAT6 Protein
Código de producto. 18748043 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
0.05 miligramo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
18748043 50 μg 0.05 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 18748043 Proveedor Novus Biologicals™ N.º de proveedor NBP1990450.05mg
Please to purchase this item. Need a web account? Register with us today!
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-95 of Mycobacterium tuberculosis Mycobacterium Tuberculosis ESAT6 The Recombinant Bacteria Mycobacterium Tuberculosis ESAT6 Protein is derived from E. coli. The Recombinant Bacteria Mycobacterium Tuberculosis ESAT6 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 50 μg