Learn More
Thermo Scientific™ Lambda DNA/HindIII Marker
Perform sizing and approximate quantification of large double-stranded DNA fragments in agarose gels with this conventional marker composed of 8 sharp bands.
Brand: Thermo Scientific™ SM0101
52.00 EUR valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Lambda DNA/HindIII Marker is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientitfic restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
The marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively, and it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction.
Highlights
- Sizing and approximate quantification of large DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
Specifications
Agarose | |
No | |
5 x 50 μg |
DNA/Hind III Marker | |
Agarose Gels |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Campaña de investigación contra el cáncer
Ahorre a lo grande en productos esenciales para la investigación contra el cáncerPromo Code: 24147Expires: 2025-07-31
-
Invitrogen
,
ABI Applied Biosystems
,
Thermo Scientific
,
Affymetrix eBioscience
Fomente su progreso
Ahorros exclusivos para que su investigación prospere.Promo Code: 24108Expires: 2025-07-04
Days
14Hours
17Minutes
54Seconds
41 -
Multiple Manufacturers
3 por 2
Ahorro exclusivo, oferta por tiempo limitado.Promo Code: 24112Expires: 2025-06-20
Days
0Hours
17Minutes
54Seconds
41New Offer Ahorrar ahora
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.