CAS RN 95809-78-2
IUPAC Nombre:
6,8-bis(benzylsulfanyl)octanoic acid
Sinónimos:
6,8-bis(benzylsulfanyl)octanoic acid
Peso molecular (g/mol):
388.58
Fórmula molecular:
C22H28O2S2
InChi Key:
ZYRLHJIMTROTBO-UHFFFAOYNA-N
SMILES:
OC(=O)CCCCC(CCSCC1=CC=CC=C1)SCC1=CC=CC=C1
1
–
5
de
5
resultados
1
CPI 613, TRC
CAS: 95809-78-2 Fórmula molecular: C22H28O2S2 Peso molecular (g/mol): 388.59 Sinónimo: 6,8-Bis[(phenylmethyl)thio]-octanoic Acid,6,8-Bis(benzylthio)-octanoic Acid Nombre IUPAC: 6,8-bis(benzilsulfanil)ácido octanoico SMILES: OC(=O)CCCCC(CCSCc1ccccc1)SCc2ccccc2
2
Tocris Bioscience™ 6,8-Bis(benzylthio)octanoic acid
PDH and KGDH inhibitor
3
Devimistat, MedChemExpress
MedChemExpress Devimistat (CPI-613) is a mitochondrial metabolism inhibitor. Devimistat is a lipoic acid antagonist that abrogates mitochondrial energy metabolism to induce apoptosis in various cancer cells.
4
5