CAS RN 905586-69-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AZ960, MedChemExpress
MedChemExpress AZ960 is a potent and specific inhibitor of the JAK2 kinase with a Ki of 0.45 nM.