CAS RN 900305-37-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
PFI-4, MedChemExpress
MedChemExpress PFI-4 is a potent and selective and cell permeable BRPF1 bromodomain inhibitor (IC50 = 80 nM).
2
Tocris Bioscience™ PFI 4
Potent and selective BRPF1 bromodomain inhibitor
3
4