Learn More
Tocris Bioscience™ PFI 4
Potent and selective BRPF1 bromodomain inhibitor
Marca: Tocris Bioscience™ 5576/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PFI 4 is a potent and selective BRPF1B bromodomain inhibitor (IC50 = 172 nM). Exhibits >100-fold selectivity for BRPF1 over a panel of other bromodomains including BRPF2 (BRD1), BRPF3 and BRD4. Disrupts BRPF1 binding to histone H3.3. Cell permeable. Inhibits osteoclastogenesis.Especificaciones
| PFI 4 | |
| N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-pyrrolidinyl)-1H-benzimidazol-5-yl]-2-methoxybenzamide | |
| Soluble to 50mM in DMSO and to 10mM in ethanol | |
| Bioactive Small Molecule |
| 900305-37-5 | |
| 50 mg | |
| Solid |
Título del producto
Research Use Only. Not For Human or Veterinary Use