CAS RN 875446-37-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Anacetrapib, MedChemExpress
MedChemExpress Anacetrapib is a potent CETP inhibitor, with IC50s of 7.9±2.5 nM and 11.8±1.9 nM for rhCETP and C13S CETP mutant, respectively.