CAS RN 847728-01-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CJ-42794, MedChemExpress
MedChemExpress CJ-42794 is a selective prostaglandin E receptor subtype 4 (EP4) antagonist, inhibits [3H]-PGE2 binding to the human EP4 receptor with a mean pKi of 8.5, a binding affinity that was at least 200-fold more selective for the human EP4 receptor than other human EP receptor subtypes (EP1, EP2, and EP3).