CAS RN 77191-36-7
IUPAC Nombre:
N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide
Sinónimos:
nefiracetam
Peso molecular (g/mol):
246.31
Fórmula molecular:
C14H18N2O2
InChi Key:
NGHTXZCKLWZPGK-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(C)=C1NC(=O)CN1CCCC1=O
1
–
4
de
4
resultados
4
Nefiracetam, MedChemExpress
MedChemExpress Nefiracetam is a GABAergic, cholinergic, and monoaminergic neuronal systems enhancer for Ro 5-4864-induced convulsions.