CAS RN 722544-51-6
IUPAC Nombre:
2-{3-[(7-{3-[ethyl(2-hydroxyethyl)amino]propoxy}quinazolin-4-yl)amino]-1H-pyrazol-5-yl}-N-(3-fluorophenyl)acetamide
Sinónimos:
2-{5-[(7-{3-[ethyl(2-hydroxyethyl)amino]propoxy}quinazolin-4-yl)amino]-2H-pyrazol-3-yl}-N-(3-fluorophenyl)acetamide
Peso molecular (g/mol):
507.57
Fórmula molecular:
C26H30FN7O3
InChi Key:
QYZOGCMHVIGURT-UHFFFAOYSA-N
SMILES:
CCN(CCO)CCCOC1=CC=C2C(NC3=NNC(CC(=O)NC4=CC(F)=CC=C4)=C3)=NC=NC2=C1
1
–
2
de
2
resultados
2
Barasertib-HQPA, MedChemExpress
MedChemExpress Barasertib-HQPA (AZD2811) is a highly selective Aurora B inhibitor with an IC50 of 0.37 nM in a cell-free assay. Barasertib-HQPA (AZD2811) induces growth arrest and apoptosis in cancer cells.