CAS RN 71441-28-6
IUPAC Nombre:
4-[(1E)-2-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)prop-1-en-1-yl]benzoic acid
Sinónimos:
ttnpb arotinoid acid tocris-0761 arotinoic acid arotinoid ro-137410 T3757_sigma
Peso molecular (g/mol):
348.49
Fórmula molecular:
C24H28O2
InChi Key:
FOIVPCKZDPCJJY-JQIJEIRASA-N
SMILES:
C\C(=C/C1=CC=C(C=C1)C(O)=O)C1=CC=C2C(=C1)C(C)(C)CCC2(C)C
1
–
3
de
3
resultados
1
Tocris Bioscience™ TTNPB
CAS: 71441-28-6 Fórmula molecular: C24H28O2
2
TTNPB, MedChemExpress
MedChemExpress TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively.
3