CAS RN 646995-35-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ BAY 58-2667 hydrochloride
Potent soluble guanylyl cyclase (sGC) activator
2
Cinaciguat hydrochloride, MedChemExpress
MedChemExpress Cinaciguat hydrochloride is a potent soluble guanylate cyclase (GC) activator with EC50 of 15 nM in platelets.
3