CAS RN 627536-09-8
IUPAC Nombre:
2-(5-chloro-2-fluorophenyl)-N-(pyridin-4-yl)pteridin-4-amine
Sinónimos:
2-(5-chloro-2-fluorophenyl)-N-(pyridin-4-yl)pteridin-4-amine
Peso molecular (g/mol):
352.76
Fórmula molecular:
C17H10ClFN6
InChi Key:
BERLXWPRSBJFHO-UHFFFAOYSA-N
SMILES:
FC1=C(C=C(Cl)C=C1)C1=NC(NC2=CC=NC=C2)=C2N=CC=NC2=N1
1
–
2
de
2
resultados
1
Tocris Bioscience™ SD 208
CAS: 627536-09-8 Fórmula molecular: C17H10ClFN6
2
SD-208, MedChemExpress
MedChemExpress SD-208 is a selective TGF-βRI (ALK5) inhibitor with IC50 of 48 nM, and > 100-fold selectivity over TGF-βRII.