CAS RN 56287-74-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
2
Afloqualone, MedChemExpress
MedChemExpress Afloqualone (HQ-495) is a GABAergic agent and has agonist activity at the β subtype of the GABAα receptor. Afloqualone has antivertiginous effects thought to be attributable to the increased sensitivity of GABA receptors of the LVN neuron site.