CAS RN 522629-08-9
IUPAC Nombre:
N3-(4-fluorophenyl)-2H-pyrazolo[3,4-d]pyrimidine-3,4-diamine
Sinónimos:
N3-(4-fluorophenyl)-2H-pyrazolo[3,4-d]pyrimidine-3,4-diamine
Peso molecular (g/mol):
244.23
Fórmula molecular:
C11H9FN6
InChi Key:
UQPMANVRZYYQMD-UHFFFAOYSA-N
SMILES:
NC1=NC=NC2=NNC(NC3=CC=C(F)C=C3)=C12
1
–
2
de
2
resultados
1
Tocris Bioscience™ CGP 57380
CAS: 522629-08-9 Fórmula molecular: C11H9FN6
2
CGP 57380, MedChemExpress
MedChemExpress CGP 57380 is a cell-permeable pyrazolo-pyrimidine compound that acts as a selective inhibitor of Mnk1 with IC50 of 2.2 μM, but has no inhibitory activity against p38, JNK1, ERK1/2, PKC, or Src-like kinases.