CAS RN 492-08-0
IUPAC Nombre:
(1R,2R,9R,10S)-7,15-diazatetracyclo[7.7.1.0²,⁷.0¹⁰,¹⁵]heptadecane
Sinónimos:
L-sparteine
Peso molecular (g/mol):
234.39
Fórmula molecular:
C15H26N2
InChi Key:
SLRCCWJSBJZJBV-TUVASFSCSA-N
SMILES:
C1CCN2C[C@H]3C[C@H](CN4CCCC[C@H]34)[C@@H]2C1
1
–
2
de
2
resultados
1
(+)-Sparteine, TRC
CAS: 492-08-0 Fórmula molecular: C15 H26 N2 Peso molecular (g/mol): 234.38 Sinónimo: 7,14-Methano-2H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocine, dodecahydro-, (7R,7aR,14R,14aS)-,7,14-Methano-2H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocine, dodecahydro-, [7R-(7α,7aα,14α,14aβ)]-,Pachycarpine (6CI,8CI),(+)-Sparteine,6α,7β,9β,11β-Sparteine,D-(+)-Sparteine,D-Sparteine,d-Sparteine SMILES: C1CCN2C[C@H]3C[C@H](CN4CCCC[C@@H]34)[C@H]2C1
2
(+)-Sparteine, MedChemExpress
MedChemExpress (+)-Sparteine is a natural alkaloid acting as a ganglionic blocking agent. (+)-Sparteine competitively blocks nicotinic ACh receptor in the neurons.