CAS RN 356057-34-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ Darapladib
Potent lp-PLA2 inhibitor
2
Darapladib, MedChemExpress
MedChemExpress Darapladib is a potent inhibitor of lipoprotein-associated phospholipase A2 (Lp-PLA2) with IC50 of 0.25 nM.